Learn More
Medchemexpress LLC HY-108449 10mg Medchemexpress, WS-12 CAS:68489-09-8 Purity:>98%
SDP
Catalog No. 501968551
Encompass_Preferred
Medchemexpress, HY-108449 10mg WS-12 CAS:68489-09-8 WS-12 is an agonist of TRPM8 with an EC50 of 39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8551 Supplier Medchemexpress LLC Supplier No. HY10844910MG
Medchemexpress, HY-108449 10mg WS-12 CAS:68489-09-8 WS-12 is an agonist of TRPM8 with an EC50 of 39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.