Learn More
Enzo Life Sciences WS-12 (25mg). CAS: 68489-09-8
SDP
Catalog No. 502009120
Encompass_Preferred
Cooling agent. High-affinity TRPM8 (cold menthol receptor 1; CMR1) agonist. Has an EC50 value ~2'000 times lower than that of (-)-menthol (ALX-420-042). Alternative name: (2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexanecarboximide. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, methanol, ethyl acetate, , dimethyl formamide or acetonitrile; insoluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9120 Supplier Enzo Life Sciences Supplier No. ALX420041M025
Cooling agent. High-affinity TRPM8 (cold menthol receptor 1; CMR1) agonist. Has an EC50 value ~2'000 times lower than that of (-)-menthol (ALX-420-042). Alternative name: (2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexanecarboximide. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, methanol, ethyl acetate, , dimethyl formamide or acetonitrile; insoluble in water or DMSO. Long Term Storage: -20°C.