Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC WS-383 | 2247544-02-9 | 99.9% | 498.46 g·mol⁻1 | C18H21Cl2N9S2 | 50MG
SDP

Catalog No. 5000238161
Encompass_Preferred

WS-383 is a potent, selective, and reversible small-molecule inhibitor of the DCN1-UBC12 protein-protein interaction (IC50 = 11 nM). In cellular assays it inhibits Cul3/1 neddylation and induces accumulation of p21, p27, and NRF2. Supplied as a solid with high purity for research use.

  • Potent DCN1-UBC12 inhibitor (IC50 = 11 nM).
  • Reversible inhibition of the DCN1-UBC12 interaction.
  • Inhibits Cul3/1 neddylation in cellular assays.
  • Induces accumulation of p21, p27, and NRF2.
  • High purity suitable for research applications.
  • Solid form with defined storage recommendations.
  • Molecular weight 498.46 g·mol⁻1; formula C18H21Cl2N9S2.

Catalog No. 50-002-38161 Supplier Medchemexpress LLC Supplier No. HY126075A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WS-383 is a potent, selective, and reversible small-molecule inhibitor of the DCN1-UBC12 protein-protein interaction (IC50 = 11 nM). In cellular assays it inhibits Cul3/1 neddylation and induces accumulation of p21, p27, and NRF2. Supplied as a solid with high purity for research use.

  • Potent DCN1-UBC12 inhibitor (IC50 = 11 nM).
  • Reversible inhibition of the DCN1-UBC12 interaction.
  • Inhibits Cul3/1 neddylation in cellular assays.
  • Induces accumulation of p21, p27, and NRF2.
  • High purity suitable for research applications.
  • Solid form with defined storage recommendations.
  • Molecular weight 498.46 g·mol⁻1; formula C18H21Cl2N9S2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.