Learn More
Medchemexpress LLC WS-383 | 2247544-02-9 | 99.9% | 498.46 g/mol | C18H21Cl2N9S2 | 5MG
SDP
Catalog No. 5000239128
Encompass_Preferred
WS-383 is a small-molecule, selective, and reversible inhibitor of the DCN1-UBC12 protein-protein interaction. It has a reported IC50 of 11 nM for disrupting DCN1-UBC12 binding, inhibits Cullin3/1 neddylation in cellular assays, and leads to accumulation of p21, p27, and NRF2. Intended for research use in biochemical and cell-based studies.
  • Potent DCN1-UBC12 interaction inhibitor (reported IC50 11 nM).
  • Selective and reversible mechanism of action.
  • Inhibits Cullin3/1 neddylation in cell-based assays.
  • Induces accumulation of cell-cycle regulators p21 and p27, and NRF2.
  • Supplied in small research pack sizes suitable for assay development.
  • High reported purity (99.9%) with recommended storage at 4°C; store in solvent at -80°C for long-term stability.
Catalog No. 50-002-39128 Supplier Medchemexpress LLC Supplier No. HY126075A5MG
WS-383 is a small-molecule, selective, and reversible inhibitor of the DCN1-UBC12 protein-protein interaction. It has a reported IC50 of 11 nM for disrupting DCN1-UBC12 binding, inhibits Cullin3/1 neddylation in cellular assays, and leads to accumulation of p21, p27, and NRF2. Intended for research use in biochemical and cell-based studies.
  • Potent DCN1-UBC12 interaction inhibitor (reported IC50 11 nM).
  • Selective and reversible mechanism of action.
  • Inhibits Cullin3/1 neddylation in cell-based assays.
  • Induces accumulation of cell-cycle regulators p21 and p27, and NRF2.
  • Supplied in small research pack sizes suitable for assay development.
  • High reported purity (99.9%) with recommended storage at 4°C; store in solvent at -80°C for long-term stability.