Learn More
Medchemexpress LLC Ws-383 | 2247544-02-9 | MFCD32182557 | 99.9% | 498.46 | C18H21Cl2N9S2 | 10MG
SDP
Catalog No. 5000235637
Encompass_Preferred
WS-383 is a potent, selective, and reversible small-molecule inhibitor of the DCN1-UBC12 protein-protein interaction (IC50 = 11 nM). It inhibits Cul3/1 neddylation in cells, resulting in accumulation of cell-cycle regulators p21 and p27 and increased NRF2, and shows cellular activity in relevant cancer cell lines.
  • Potent and selective inhibitor with low-nanomolar biochemical potency.
  • Cellular inhibition of Cul3 neddylation at low micromolar concentrations.
  • Induces accumulation of p21, p27, and NRF2 in treated cells.
  • High-purity solid suitable for biochemical and cellular assays.
  • Available as solid and as a DMSO solution for cell-based work.
Catalog No. 50-002-35637 Supplier Medchemexpress LLC Supplier No. HY126075A10MG
Add to Cart
Add to Cart
WS-383 is a potent, selective, and reversible small-molecule inhibitor of the DCN1-UBC12 protein-protein interaction (IC50 = 11 nM). It inhibits Cul3/1 neddylation in cells, resulting in accumulation of cell-cycle regulators p21 and p27 and increased NRF2, and shows cellular activity in relevant cancer cell lines.
  • Potent and selective inhibitor with low-nanomolar biochemical potency.
  • Cellular inhibition of Cul3 neddylation at low micromolar concentrations.
  • Induces accumulation of p21, p27, and NRF2 in treated cells.
  • High-purity solid suitable for biochemical and cellular assays.
  • Available as solid and as a DMSO solution for cell-based work.