Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ws-917 | 10 MG
SDP

Catalog No. 5000295259
Encompass_Preferred

WS-917 is a small-molecule research compound characterized as a potent, orally active ABCB1 (P-glycoprotein) modulator. It enhances ABCB1 ATPase activity, promotes CD8+ T-cell activation, and has been shown to potentiate the antitumor effect of paclitaxel when combined with anti-PD-1 therapy. Intended for research use; follow appropriate storage and handling.

  • Potent, orally active ABCB1 modulator.
  • Enhances ABCB1 ATPase activity.
  • Promotes CD8+ T-cell activation.
  • Shown to enhance paclitaxel antitumor effect in combination with anti-PD-1 therapy.
  • Molecular formula C16H13N5OS2; molecular weight 355.44 g/mol.
  • Recommended storage: 4°C under nitrogen; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) under nitrogen.

Catalog No. 50-002-95259 Supplier Medchemexpress LLC Supplier No. HY16852810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WS-917 is a small-molecule research compound characterized as a potent, orally active ABCB1 (P-glycoprotein) modulator. It enhances ABCB1 ATPase activity, promotes CD8+ T-cell activation, and has been shown to potentiate the antitumor effect of paclitaxel when combined with anti-PD-1 therapy. Intended for research use; follow appropriate storage and handling.

  • Potent, orally active ABCB1 modulator.
  • Enhances ABCB1 ATPase activity.
  • Promotes CD8+ T-cell activation.
  • Shown to enhance paclitaxel antitumor effect in combination with anti-PD-1 therapy.
  • Molecular formula C16H13N5OS2; molecular weight 355.44 g/mol.
  • Recommended storage: 4°C under nitrogen; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.