Learn More
SPEX CERTIPREP LLC 3P WS - UV254 Absorbance
Catalog No. NC9801293
Encompass_Preferred
(NSI by Zeptometrix) A 21 mL concentrate for determination of UV254 absorbance. Formulated in the NELAC range of 0.05 - 0.7 cm(-1).
Catalog No. NC9801293 Supplier SPEX CERTIPREP LLC 3P Supplier No. PEI085
(NSI by Zeptometrix) A 21 mL concentrate for determination of UV254 absorbance. Formulated in the NELAC range of 0.05 - 0.7 cm(-1).