Learn More
TARGETMOL CHEMICALS INC WS3 25MG
SDP
Catalog No. 502577238
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WS3 a (beta) cell proliferation inducer regulates Erb3 binding protein-1 (EBP1) and the I(kappa)B kinase pathway. purity: 98%
Catalog No. 50-257-7238 Supplier TARGETMOL CHEMICALS INC Supplier No. T205525MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WS3 a (beta) cell proliferation inducer regulates Erb3 binding protein-1 (EBP1) and the I(kappa)B kinase pathway. purity: 98%