Learn More
Medchemexpress LLC HY-137605 5mg Medchemexpress, WSF1-IN-1 CAS:2379577-82-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1376055MG
Encompass_Preferred
Medchemexpress, HY-137605 5mg WSF1-IN-1 CAS:2379577-82-7 WSF1-IN-1 (compound 136), an orally active WSF1 inhibitor, can be used in the study for WSF1 (Wolfram syndrome) related tumors, with IC50 values of 0.33 μM and >27 μM in HepG2 parental and HepG2 WFS1 KO cell lines, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1503