Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WSFA-2, 0.1 wt. Each Component in Deionized Water, Qualitative Standard, MilliporeSigma™ Supelco™

Catalog No. 111004252 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4252 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4252 Supplier MilliporeSigma Supelco Supplier No. 47056
Add to Cart
Add to Cart
WSFA-2 may be used for identifying and quantifying purgeable halocarbons in municipal and industrial wastewater in accordance with EPA 624 method. The pre-requisite of EPA 624 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry. WSFA-2 may also be utilized for the qualitative determination of free fatty acids in aqueous solutions. It is recommended that it should be purchased from chemical manufacturers with knowledge in the preparation, handling, storage, and shipment of volatile analytes.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 wt.% each component in deionized water
Quantity 5 mL
Grade Qualitative Standard
Concentration or Composition (by Analyte or Components) Acetic acid; Butyric acid; Isobutyric acid; Isovaleric acid; Propionic acid; Valeric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.