Learn More
WSFA-4, 0.1 wt. Each Component in Deionized Water, Qualitative Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Concentration | 0.1 wt.% each component in deionized water |
| Quantity | 5 mL |
| Grade | Qualitative Standard |
| Concentration or Composition (by Analyte or Components) | Acetic acid; Butyric acid; Isobutyric acid; Isovaleric acid; 2-Methylbutyric acid; Propionic acid; Valeric acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.