Learn More
Electron Microscopy Sciences Wash-Off™ Wash Off Label Plain, 4.00" x 1.00"
SDP
Catalog No. 501899650
Encompass_Preferred
Rinse away leaving no adhesive residue. Labels dissolve under running water in less than 30 seconds. Labels Wash-Off in 1 rinse cycle at any water temperature. 100% biodegradable and FDA compliant. Labels resist exposure to steam at 121 degree C, however a build-up of liquid water on the label surface will dissolve the material. Long term storage best between 32 degree F to 70 degree F (0 degree C to 21 degree C). Accepts laser printing and other markings containers.
Size: 4.00" x 1.00"
Catalog No. 50-189-9650 Supplier Electron Microscopy Sciences Supplier No. 7182620
Rinse away leaving no adhesive residue. Labels dissolve under running water in less than 30 seconds. Labels Wash-Off in 1 rinse cycle at any water temperature. 100% biodegradable and FDA compliant. Labels resist exposure to steam at 121 degree C, however a build-up of liquid water on the label surface will dissolve the material. Long term storage best between 32 degree F to 70 degree F (0 degree C to 21 degree C). Accepts laser printing and other markings containers.
Size: 4.00" x 1.00"