Learn More
AdipoGen WSP1
SDP
Supplier:  AdipoGen CDXW0001M005
Encompass_Preferred
Chemical. CAS 1352750-34-5. Formula C33H21NO6S2. MW 591.65. Synthetic Hydrogen sulfide H2S is an important gaseous mediator, like nitric oxide, that has significant effects on the immunological, neurological, cardiovascular and pulmonary systems of mammals. WSP-1 is a reactive disulfide-containing, fluorescent probe designed to detect H2S in biological samples and cells. Through a reaction-based fluorescent turn-on strategy, WSP-1 selectively and rapidly reacts with H2S to generate benzodithiolone and a fluorophore with excitation and emission maxima of 465 and 515 nm, respectively.
Catalog No. 50-255-9909