Learn More
Medchemexpress LLC WST-1 | 150849-52-8 | 97.3% | 651.34 | 50 MG
SDP
Catalog No. 5000348074
Encompass_Preferred
WST-1 is a water-soluble tetrazolium salt that induces intracellular mitochondrial dehydrogenase to conduct an NADH-dependent enzyme digestion reaction, releasing a water-soluble methyl benzene product. It can be used for the detection of cell proliferation and cytotoxicity by determining the light absorption value at 450 nm. This product is for research use only.
  • Water-soluble tetrazolium salt
  • Induces intracellular mitochondrial dehydrogenase activity
  • Releases a water-soluble methyl benzene product
  • Suitable for detection of cell proliferation
  • Suitable for cytotoxicity analysis
  • Determines light absorption value at 450 nm
  • Used in cell staining analysis
  • Relevant for cell function analysis
  • Considered a cell viability dye
Catalog No. 50-003-48074 Supplier Medchemexpress LLC Supplier No. HY13697650MG
WST-1 is a water-soluble tetrazolium salt that induces intracellular mitochondrial dehydrogenase to conduct an NADH-dependent enzyme digestion reaction, releasing a water-soluble methyl benzene product. It can be used for the detection of cell proliferation and cytotoxicity by determining the light absorption value at 450 nm. This product is for research use only.
  • Water-soluble tetrazolium salt
  • Induces intracellular mitochondrial dehydrogenase activity
  • Releases a water-soluble methyl benzene product
  • Suitable for detection of cell proliferation
  • Suitable for cytotoxicity analysis
  • Determines light absorption value at 450 nm
  • Used in cell staining analysis
  • Relevant for cell function analysis
  • Considered a cell viability dye