Learn More
Medchemexpress LLC WST-3 | 515111-36-1 | 99.6% | 696.34 g/mol | C19H10IN6NaO10S2 | 25MG
SDP
Catalog No. 5000238236
Encompass_Preferred
WST-3 is a water-soluble tetrazolium dye used as a colorimetric electron-transfer indicator in NADH-dependent assays. In the presence of an electron mediator, it is reduced by NADH to a water-soluble formazan that has an absorption maximum at 433 nm, allowing spectrophotometric quantification of reducing equivalents in cell viability and enzyme activity assays.
  • Reduces to a water-soluble formazan in the presence of NADH and an electron mediator.
  • Formazan product absorbs at 433 nm for spectrophotometric detection.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO at 100 mg/mL; aqueous formulations are achievable using co-solvents.
  • Store solid protected from moisture and light at 4°C.
Catalog No. 50-002-38236 Supplier Medchemexpress LLC Supplier No. HY12592125MG
WST-3 is a water-soluble tetrazolium dye used as a colorimetric electron-transfer indicator in NADH-dependent assays. In the presence of an electron mediator, it is reduced by NADH to a water-soluble formazan that has an absorption maximum at 433 nm, allowing spectrophotometric quantification of reducing equivalents in cell viability and enzyme activity assays.
  • Reduces to a water-soluble formazan in the presence of NADH and an electron mediator.
  • Formazan product absorbs at 433 nm for spectrophotometric detection.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO at 100 mg/mL; aqueous formulations are achievable using co-solvents.
  • Store solid protected from moisture and light at 4°C.