Learn More
Medchemexpress LLC WST-3 (water-soluble tetrazolium dye) | 515111-36-1 | 99.6% | 696.34 g/mol | C19H10IN6NaO10S2 | 10MG
SDP
Catalog No. 5000237845
Encompass_Preferred
WST-3 is a water-soluble tetrazolium dye used to measure cellular metabolic activity and NADH-dependent enzymatic reactions. In the presence of an electron mediator, WST-3 is reduced by NADH to a water-soluble formazan with an absorption maximum near 433 nm, enabling sensitive colorimetric and fluorometric detection for viability and enzyme assays.
  • Produces water-soluble formazan detectable at ~433 nm.
  • Suitable for cell viability and dehydrogenase activity assays.
  • Compatible with electron mediators for NADH-dependent reduction.
  • Soluble in DMSO; examples of in vivo formulation are available.
  • High purity (99.6%).
  • Supplied in small research-scale quantities for assay development.
Catalog No. 50-002-37845 Supplier Medchemexpress LLC Supplier No. HY12592110MG
WST-3 is a water-soluble tetrazolium dye used to measure cellular metabolic activity and NADH-dependent enzymatic reactions. In the presence of an electron mediator, WST-3 is reduced by NADH to a water-soluble formazan with an absorption maximum near 433 nm, enabling sensitive colorimetric and fluorometric detection for viability and enzyme assays.
  • Produces water-soluble formazan detectable at ~433 nm.
  • Suitable for cell viability and dehydrogenase activity assays.
  • Compatible with electron mediators for NADH-dependent reduction.
  • Soluble in DMSO; examples of in vivo formulation are available.
  • High purity (99.6%).
  • Supplied in small research-scale quantities for assay development.