Learn More
Medchemexpress LLC HY-D0831 200mg , WST-8 CAS:193149-74-5 Purity:98%
SDP
Catalog No. 501492615
Encompass_Preferred
Medchemexpress, HY-D0831 200mg WST-8 CAS:193149-74-5 Formula:C20H14N6NaO11S2 Purity:98% WST-8 is a water-soluble tetrazolium dye, WST-8 enhances sensitivity of the WST-8-based assay over the conventional MTS-based assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2615 Supplier Medchemexpress LLC Supplier No. HYD0831200MG
Medchemexpress, HY-D0831 200mg WST-8 CAS:193149-74-5 Formula:C20H14N6NaO11S2 Purity:98% WST-8 is a water-soluble tetrazolium dye, WST-8 enhances sensitivity of the WST-8-based assay over the conventional MTS-based assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.