Learn More
Selleck Chemical LLC WST-8
SDP
Catalog No. 5000001061
Encompass_Preferred
WST-8 is a water-soluble tetrazolium salt used for assessing cell metabolic activity that produces corresponding formazan dye that absorbs at 460 nm WST-8 is typically used as a cell viability indicator in cell proliferation assays
Catalog No. 50-000-01061 Supplier Selleck Chemical LLC Supplier No. S7764100MG
WST-8 is a water-soluble tetrazolium salt used for assessing cell metabolic activity that produces corresponding formazan dye that absorbs at 460 nm WST-8 is typically used as a cell viability indicator in cell proliferation assays