Learn More
TRIBIOSCIENCE INC WST-8 or Cell Count Kit-8 (CCK-8), 500 Tests
SDP
Catalog No. NC2886949
Encompass_Preferred
Cell Counting Kit-8 (CCK-8) can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.
Catalog No. NC2886949 Supplier TRIBIOSCIENCE INC Supplier No. TBS2022500
Cell Counting Kit-8 (CCK-8) can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.