Learn More
Cayman Chemical WST-1 Cell Proliferation Assay Kit, 5 ea
SDP
Catalog No. 502886797
Encompass_Preferred
Cayman’s WST-1 Proliferation Assay is based on the reduction of tetrazolium salt WST-1 to a soluble formazan dye by electron transport across the plasma membrane of dividing cells. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation.
Catalog No. 50-288-6797 Supplier Cayman Chemical Supplier No. 100088835 EA
Cayman’s WST-1 Proliferation Assay is based on the reduction of tetrazolium salt WST-1 to a soluble formazan dye by electron transport across the plasma membrane of dividing cells. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation.