Learn More
New England Biolabs, Inc. WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG)- 500 rxn
SDP
Catalog No. 502229440
Encompass
The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is designed to provide a simple one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase. This mix contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution. The inclusion of thermolabile UDG prevents carryover contamination where unintended product of a previous amplification can serve as the substrate of a subsequent reaction. The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is compatible with multiple detection methods including turbidity detection real-time fluorescence detection and end-point visualization.
This item is not returnable. View return policy
Catalog No. 50-222-9440 Supplier New England Biolabs, Inc. Supplier No. M1708L
The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is designed to provide a simple one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase. This mix contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution. The inclusion of thermolabile UDG prevents carryover contamination where unintended product of a previous amplification can serve as the substrate of a subsequent reaction. The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is compatible with multiple detection methods including turbidity detection real-time fluorescence detection and end-point visualization.