Learn More
Medchemexpress LLC HY-100871 5mg Medchemexpress, WT-161 CAS:1206731-57-8 Purity:>98%
SDP
Catalog No. 501931978
Encompass_Preferred
Medchemexpress, HY-100871 5mg WT-161 CAS:1206731-57-8 WT-161 is a potent and selective HDAC6 inhibitor with an IC 50 of 0.40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1978 Supplier Medchemexpress LLC Supplier No. HY1008715MG
Medchemexpress, HY-100871 5mg WT-161 CAS:1206731-57-8 WT-161 is a potent and selective HDAC6 inhibitor with an IC 50 of 0.40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.