Learn More
ATCC WT 9-7; Kidney; Human (Homo sapiens)
Supplier:  ATCC CRL2830
Encompass
WT 9-7is an epithelial cell line that was isolated in 1999 from the proximal tubule of an autosomal dominant polycystic kidney disease ADPKD female patient. This cell line was deposited by M Loghman-Adham.
Catalog No. 50-238-3191