Learn More
ATCC Wt MEFs; Mouse Embryo Fibroblast; Mouse (Mus musculus)
Supplier:  ATCC CRL2991
Encompass
Wt MEFs is a mouse fibroblast cell line isolated in 2000 and can be used as a control to study the role of mitochondrial fusion in cell physiology. MEFs were derived from embryos 10.5 days gestation.
Catalog No. 50-238-3816
 Restricted Item