Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7490MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: WT1/857. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. Cellular Staining: Nuclear. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human WT1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 47-55kDa.Chromosome: 11p13. Gene Symbol: WT1. Gene ID: 7490. Ensemble: ENSG00000184937. Uniprot: NP_001185480.
Catalog No. 50-188-5287