Learn More
Aviva Systems Biology Corp WT1 Antibody - middle region (ARP37989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WT1. Alternate names: GUD, WAGR, WIT-2, WT33, AWT1, NPHS4, EWS-WT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56476