Learn More
Aviva Systems Biology Corp WT1 Antibody - N-terminal region (ARP37988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human WT1. Alternate names: AWT1, GUD, WAGR, WIT-2, WT33, NPHS4, EWS-WT1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56475