Learn More
Cole-Parmer Stuart Digital Water Bath, stainless steel, 15 L, 230 VAC
Supplier:  Cole-Parmer 1212271
Encompass_Preferred
These utility baths feature a clear LED temperature display with an analog dial, making set point adjustments quick and ensuring accurate temperature control. Polycarbonate lid provides full view of bath contents and reduces heat loss. Low water level sensor prevents dry boiling, and overtemperature protection system monitors temperature and controls heater in the event of a fault.
Built-in drain empties stainless steel tank for water changes and maintenance. The heater is mounted beneath the tank, so it is not in the way.
Catalog No. 50-196-0427