Learn More
Growcells Water for CO2 Incubators 6 x 2L Bottles
Catalog No. 501535069
Encompass_Preferred
Growcells Water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers
Catalog No. 50-153-5069 Supplier Growcells Supplier No. ISDW20006
Growcells Water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers