Learn More
Growcells WATER FOR CO2 INCUBATORS 2 X 2L BOTTLES
Catalog No. 501535068
Encompass_Preferred
Growcells Water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers
Catalog No. 50-153-5068 Supplier Growcells Supplier No. ISDW20002
Growcells Water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers