Learn More
Fine Science Tools Fine Science Tools Wullstein Retractor, Stainless Steel, 3x3 Blunt Teeth, 11 cm, 1 ea
SDP
Catalog No. NC3917981
Encompass
The Wullstein retractor is a specialized surgical instrument designed for tissue retraction. Constructed from durable surgical stainless steel (Inox), this instrument is engineered for precision and reliability in laboratory research environments.
  • Manufactured from surgical stainless steel for long-term durability
  • Equipped with 3x3 blunt teeth for effective tissue engagement
  • Designed with a total length of 11 cm
  • Features angled-down geometry for improved access
  • Teeth dimensions measure 16 mm in length and 2 mm in width
  • Lightweight design weighing approximately 49 g
  • Originates from Germany
Catalog No. NC3917981 Supplier Fine Science Tools Supplier No. 1701811
The Wullstein retractor is a specialized surgical instrument designed for tissue retraction. Constructed from durable surgical stainless steel (Inox), this instrument is engineered for precision and reliability in laboratory research environments.
  • Manufactured from surgical stainless steel for long-term durability
  • Equipped with 3x3 blunt teeth for effective tissue engagement
  • Designed with a total length of 11 cm
  • Features angled-down geometry for improved access
  • Teeth dimensions measure 16 mm in length and 2 mm in width
  • Lightweight design weighing approximately 49 g
  • Originates from Germany