Learn More
Enzo Life Sciences WESTERNVIEW Detection Kit (Anti-Rabbit) (1 Kit)
SDP
Catalog No. 502013861
Encompass_Preferred
The WESTERNVIEW™ Detection Kit (Anti-Rabbit) can be used to identify primary antibodies on a Western Blot using immunodetection. The kit contains a complete reagent set, with optimized ready-to-use or ready-to-dilute reagents. Assay Time: 2 hours. Applications: WB, Colorimetric detection . Species reactivity: Rabbit.
Catalog No. 50-201-3861 Supplier Enzo Life Sciences Supplier No. ENZKIT1830001
The WESTERNVIEW™ Detection Kit (Anti-Rabbit) can be used to identify primary antibodies on a Western Blot using immunodetection. The kit contains a complete reagent set, with optimized ready-to-use or ready-to-dilute reagents. Assay Time: 2 hours. Applications: WB, Colorimetric detection . Species reactivity: Rabbit.