Learn More
Apexbio Technology LLC WWL 70 947669-91-2 10mg
SDP
Supplier:  Apexbio Technology LLC A889110
Encompass_Preferred
WWL 70 (CAS 947669-91-2) is a small molecule inhibitor targeting / hydrolase domain-containing protein 6 (ABHD6) a serine hydrolase implicated in lipid metabolism Developed as an O-arylated derivative of carbamate 18 WWL 70 demonstrates approximately five-fold enhanced potency exhibiting an IC50 of 70 nM for ABHD6 inhibition Activity-based protein profiling and proteomic studies indicate that WWL 70 selectively inhibits ABHD6 over a panel of 27 serine hydrolases with minimal activity against ABHD10 at concentrations up to 100 M WWL 70 serves as a valuable chemical probe for elucidating ABHD6 function and investigating lipid signaling pathways in neurological research
Catalog No. 50-000-95978