Learn More
TARGETMOL CHEMICALS INC WWL123 5MG
SDP
Catalog No. 502580971
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. WWL123 is a potent and selective ABHD6 inhibitor (IC50=430 nM). WWL123 crosses the blood-brain-barrier and inhibits ABHD6 in brain parenchyma. ABHD6 blockade by WWL123 exerts an antiepileptic effect in Pentylenetetrazole (PTZ)-induced epileptiform seizures and spontaneous seizures in R6/2 mice. purity: 99%
Catalog No. 50-258-0971 Supplier TARGETMOL CHEMICALS INC Supplier No. T221715MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. WWL123 is a potent and selective ABHD6 inhibitor (IC50=430 nM). WWL123 crosses the blood-brain-barrier and inhibits ABHD6 in brain parenchyma. ABHD6 blockade by WWL123 exerts an antiepileptic effect in Pentylenetetrazole (PTZ)-induced epileptiform seizures and spontaneous seizures in R6/2 mice. purity: 99%