Learn More
Aviva Systems Biology Corp WWP1 Antibody - N-terminal region (ARP43084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WWP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WWP1. Alternate names: AIP5, DKFZp434D2111, Tiul1, hSDRP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58420