Learn More
Aviva Systems Biology Corp WWP2 Antibody - C-terminal region (ARP43089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WWP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human WWP2. Alternate names: AIP2, WWp2-likeApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58424