Learn More
Aviva Systems Biology Corp WWP2 Antibody - middle region (ARP43088_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43088_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WWP2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human WWP2. Alternate names: AIP2, WWp2-likeApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58423