Learn More
Waters Corp Waters wwPTFE Acrodisc Syringe Filter, 0.45 μm, 13 mm, 1000/Pk
Supplier:  Waters Corp 186009319
Encompass_Preferred
This product features water wettable, hydrophilic membranes that are designed for use with aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions. It is characterized by low protein binding and minimal levels of UV-absorbing extractables.
  • All-purpose hydrophilic membrane
  • Compatible with a wide range of solutions, including aqueous, acidic, basic, and organic
  • Low protein binding
  • Low levels of UV-absorbing extractables
Catalog No. 50-203-8979