Learn More
Waters Corp Syringe Filter, Acrodisc PSF, wwPTFE, 0.45 μm, 25 mm
Supplier:  Waters Corp 186009328
Encompass_Preferred
This syringe filter features a water wettable, hydrophilic, polytetrafluoroethene (wwPTFE) membrane, described as a next generation, all-purpose, hydrophilic membrane suitable for aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions. It is designed for HPLC/UHPLC sample preparation, offering low protein binding, low levels of UV-absorbing extractables, and extending column life.
  • Suitable for use with aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions.
  • Low protein binding.
  • Low levels of UV-absorbing extractables.
  • Extends column life up to 52 times.
  • Improves result quality with up to 24% less filter extractables and lower adsorption.
  • Allows the use of one membrane filter type for all sample preparation applications.
Catalog No. 50-239-1499