Learn More
Aviva Systems Biology Corp WWTR1 Antibody - N-terminal region (ARP32540_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32540_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WWTR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WWTR1. Alternate names: DKFZP586I1419, TAZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54643