Learn More
AdipoGen WY-14643 (50 mg)
SDP
Supplier:  AdipoGen AGCR13566M050
Encompass_Preferred
Chemical. CAS: 50892-23-4. Formula: C14H14ClN3O2S. MW: 323.8. Potent peroxisome proliferator-activated receptor (PPARalpha) activator. Activates also PPARgamma but not PPARdelta. Potent anti-hypercholesterolemic agent. Hypolipidemic compound. Lipogenesis inducer. Tumor promoter. Causes increased cell proliferation and decreased apoptosis. Anti-inflammatory. Inhibits NF-kappaB transcriptional activity and decreases the inflammatory response by reducing the production of inflammatory cytokines (TNF-alpha, IL1beta). Reduces oxidative stress. Increases fatty acid oxidation. Directly affects insulin signaling. Increases glucose uptake. Reviews.
Catalog No. 50-205-3934