Learn More
Apexbio Technology LLC WY-14643 (Pirinixic Acid), 250mg. Cas: 50892-23-4 MFCD: MFCD00191335. PPARα agonist,selective and highly potent
SDP
Catalog No. 501904912
Encompass_Preferred
WY-14643, also known as Pirinixic Acid, has an agonistic action as peroxisome proliferator-activated receptor (PPAR). It is shown that aliphatic -substitution of WY-14643 enhances both PPAR and PPAR agonism. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4912 Supplier Apexbio Technology LLC Supplier No. A4305250
WY-14643, also known as Pirinixic Acid, has an agonistic action as peroxisome proliferator-activated receptor (PPAR). It is shown that aliphatic -substitution of WY-14643 enhances both PPAR and PPAR agonism. Other sizes are available. Please inqury us for quote.