Learn More
Selleck Chemical LLC Desvenlafaxine Succinate hydrate S4112-25mg
SDP
Catalog No. 503111332
Encompass_Preferred
Desvenlafaxine Succinate hydrate (WY 45233) is a new serotonin (5-HT) transporter and norepinephrine (NE) transporter reuptake inhibitor with Ki of 40 2 nM and 558 4 nM respectively
Catalog No. 50-311-1332 Supplier Selleck Chemical LLC Supplier No. S411225MG
Desvenlafaxine Succinate hydrate (WY 45233) is a new serotonin (5-HT) transporter and norepinephrine (NE) transporter reuptake inhibitor with Ki of 40 2 nM and 558 4 nM respectively