Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wy 49051 | 113418-56-7 | 98.7% | 487.59 | C28H33N5O3 | 1 ML
SDP

Catalog No. 5000249300
Encompass_Preferred

Wy 49051 is a potent, orally active H1 histamine receptor antagonist used in pharmacological research. It inhibits H1 receptors with an IC50 of 44 nM and shows additional affinity for α1 receptors. Supplied as a solid or as a ready-to-use 10 mM solution in DMSO, the compound is intended for in vitro pharmacology and receptor binding studies.

  • Potent H1 receptor antagonist with IC50 of 44 nM.
  • High affinity for α1 receptor (IC50 ≈ 8 nM).
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • Purity 98.7%.
  • Molecular formula C28H33N5O3; molecular weight 487.59.
  • Appearance: light yellow solid.
  • Recommended storage: powder -20°C or in solvent -80°C.

Catalog No. 50-002-49300 Supplier Medchemexpress LLC Supplier No. HY10183010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wy 49051 is a potent, orally active H1 histamine receptor antagonist used in pharmacological research. It inhibits H1 receptors with an IC50 of 44 nM and shows additional affinity for α1 receptors. Supplied as a solid or as a ready-to-use 10 mM solution in DMSO, the compound is intended for in vitro pharmacology and receptor binding studies.

  • Potent H1 receptor antagonist with IC50 of 44 nM.
  • High affinity for α1 receptor (IC50 ≈ 8 nM).
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • Purity 98.7%.
  • Molecular formula C28H33N5O3; molecular weight 487.59.
  • Appearance: light yellow solid.
  • Recommended storage: powder -20°C or in solvent -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.