Learn More
Waters Corp Wyatt Technology Disposable MicroCuvette Kit, 30 Pack, NanoStar II, COC
Catalog No. NC3990022 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Disposable MicroCuvette Kit is designed for use with NanoStar II and ZetaStar instruments. It provides 30 disposable cuvettes for various laboratory applications.
  • Package of 30 disposable cuvettes with 3 reusable caps.
  • Compatible with NanoStar II and ZetaStar instruments.
  • Requires a minimum volume of 4 μL.
  • Made from cyclic olefin copolymers (COC).
  • Compatible with aqueous solvents only.
  • Temperature range of -10°C to 80°C.
Catalog No. NC3990022 Supplier Waters Corp Supplier No. WZDMC
This Disposable MicroCuvette Kit is designed for use with NanoStar II and ZetaStar instruments. It provides 30 disposable cuvettes for various laboratory applications.
  • Package of 30 disposable cuvettes with 3 reusable caps.
  • Compatible with NanoStar II and ZetaStar instruments.
  • Requires a minimum volume of 4 μL.
  • Made from cyclic olefin copolymers (COC).
  • Compatible with aqueous solvents only.
  • Temperature range of -10°C to 80°C.