Learn More
Medchemexpress LLC HY-124136 5mg Medchemexpress, WYC-209 CAS:2131803-90-0 Purity:>98%
SDP
Catalog No. 501932588
Encompass_Preferred
Medchemexpress, HY-124136 5mg WYC-209 CAS:2131803-90-0 WYC-209, a synthetic retinoid, is a retinoic acid receptor (RAR) agonist. WYC-209 induces apoptosis primarily via the caspase 3 pathway ( IC 50 =0.19 μM for inmalignant murine melanoma TRCs), and has long-term effects with little toxicity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2588 Supplier Medchemexpress LLC Supplier No. HY1241365MG
Medchemexpress, HY-124136 5mg WYC-209 CAS:2131803-90-0 WYC-209, a synthetic retinoid, is a retinoic acid receptor (RAR) agonist. WYC-209 induces apoptosis primarily via the caspase 3 pathway ( IC 50 =0.19 μM for inmalignant murine melanoma TRCs), and has long-term effects with little toxicity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.