Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wyc-210 (tazarotene derivative) | 2131803-93-3 | 99.2% | 340.40 | C18H16N2O3S | 5 MG
SDP

Catalog No. 5000222394
Encompass_Preferred

WYC-210 is a tazarotene-derivative small-molecule retinoid intended for research use. The compound contains an alkyne functional group and is used as a click-chemistry reagent; it has been reported to exhibit relatively low anticancer activity. The material is supplied with a high purity specification and explicit storage recommendations for solid and solution forms.

  • Contains an alkyne group suitable for click chemistry.
  • Tazarotene-derived retinoid scaffold.
  • High reported purity, suitable for analytical and synthetic applications.
  • Available in small-mass packages for laboratory-scale experiments.
  • Recommended storage conditions specified for powder and solutions.

Catalog No. 50-002-22394 Supplier Medchemexpress LLC Supplier No. HY1365545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WYC-210 is a tazarotene-derivative small-molecule retinoid intended for research use. The compound contains an alkyne functional group and is used as a click-chemistry reagent; it has been reported to exhibit relatively low anticancer activity. The material is supplied with a high purity specification and explicit storage recommendations for solid and solution forms.

  • Contains an alkyne group suitable for click chemistry.
  • Tazarotene-derived retinoid scaffold.
  • High reported purity, suitable for analytical and synthetic applications.
  • Available in small-mass packages for laboratory-scale experiments.
  • Recommended storage conditions specified for powder and solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.