Learn More
Selleck Chemical LLC WYE-125132 (WYE-132) S2661-5mg
SDP
Catalog No. 503110225
Encompass_Preferred
WYE-125132 (WYE-132) is a highly potent ATP-competitive mTOR inhibitor with IC50 of 0 19 nM highly selective for mTOR versus PI3Ks or PI3K-related kinases hSMG1 and ATR
Catalog No. 50-311-0225 Supplier Selleck Chemical LLC Supplier No. S26615MG
WYE-125132 (WYE-132) is a highly potent ATP-competitive mTOR inhibitor with IC50 of 0 19 nM highly selective for mTOR versus PI3Ks or PI3K-related kinases hSMG1 and ATR