Learn More
Enzo Life Sciences WYE-132 (5mg). CAS: 1144068-46-1
SDP
Catalog No. 502013440
Encompass_Preferred
A highly potent, ATP-competitive, and specific mTOR kinase inhibitor. Shows an anti-proliferative activity against a variety of tumor cell lines. WYE-132 causes inhibition of protein synthesis and cell size, induction of apoptosis, and cell cycle progression. Alternative name: 1-{4-[1-(1,4-Dioxa-spiro[4.5]dec-8-yl)-4-(8-oxa-3-aza-bicyclo[3.2.1]oct-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-6-yl]-phenyl}-3-methyl-urea, WYE-125132. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3440 Supplier Enzo Life Sciences Supplier No. ENZCHM1400005
A highly potent, ATP-competitive, and specific mTOR kinase inhibitor. Shows an anti-proliferative activity against a variety of tumor cell lines. WYE-132 causes inhibition of protein synthesis and cell size, induction of apoptosis, and cell cycle progression. Alternative name: 1-{4-[1-(1,4-Dioxa-spiro[4.5]dec-8-yl)-4-(8-oxa-3-aza-bicyclo[3.2.1]oct-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-6-yl]-phenyl}-3-methyl-urea, WYE-125132. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.