Learn More
Medchemexpress LLC Methyl-4-(6-{4-[(methoxycarbonyl)amino]phenyl}-4-morpholin-4-yl-1H-pyrazolo[3,4-d]pyrimidin-1-yl) | 1062169-56-5 | 98.0% | 5 MG
SDP
Catalog No. 501969604
Encompass_Preferred
WYE-354 is an ATP-competitive mTOR inhibitor used in research to block mTORC1 and mTORC2 signaling and to study PI3K/mTOR pathway modulation in vitro. It is supplied as a high-purity small molecule in solid form or as a ready-to-use DMSO solution for laboratory assays.
  • ATP-competitive mTOR inhibitor with IC50 ≈ 5 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 495.53 g·mol⁻¹ and formula C24H29N7O5.
  • Commonly used for pathway, signaling, and autophagy research in vitro.
Catalog No. 50-196-9604 Supplier Medchemexpress LLC Supplier No. HY120345MG
WYE-354 is an ATP-competitive mTOR inhibitor used in research to block mTORC1 and mTORC2 signaling and to study PI3K/mTOR pathway modulation in vitro. It is supplied as a high-purity small molecule in solid form or as a ready-to-use DMSO solution for laboratory assays.
  • ATP-competitive mTOR inhibitor with IC50 ≈ 5 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 495.53 g·mol⁻¹ and formula C24H29N7O5.
  • Commonly used for pathway, signaling, and autophagy research in vitro.